Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244188.1 | NP_001231117.1 | 5 - 6 | 4346 | 87 |
XM_005656279.3 | XP_005656336.1 | - | 6078 | 87 | |
XM_021097992.1 | XP_020953651.1 | - | 4756 | 87 | |
XM_021097993.1 | XP_020953652.1 | - | 4856 | 87 | |
XM_021097994.1 | XP_020953653.1 | - | 4593 | 87 | |
XM_021097995.1 | XP_020953654.1 | - | 4389 | 87 |