Assay Design



Gene Transcripts

Gene Symbol
RGCC
Entrez Gene ID
100625944
Gene Name
regulator of cell cycle
Gene Aliases
RGC32
Chromosome Location
Chr.11: 25481845 - 25496548 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244227.1 NP_001231156.1 4 - 5 484 109
GenBank mRNA AK392471.1 - 4 - 5 582 109
AY610446.1 - 4 - 5 484 109

View Full Product Details