Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244282.1 | NP_001231211.1 | 4 - 5 | 1206 | 115 |
GenBank mRNA | AK235689.1 | - | 4 - 5 | 1194 | 115 |
AK395562.1 | - | 4 - 5 | 1206 | 115 | |
FS663292.1 | - | 2 - 3 | 617 | 115 |