Assay Design



Gene Transcripts

Gene Symbol
PPID
Entrez Gene ID
100515957
Gene Name
peptidylprolyl isomerase D
Gene Aliases
-
Chromosome Location
Chr.8: 47656269 - 47702805 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244386.1 NP_001231315.1 7 - 8 972 94
XM_021100359.1 XP_020956018.1 - 1571 94
GenBank mRNA AK344118.1 - 7 - 8 967 94
AY609657.1 - 7 - 8 972 94

View Full Product Details