Assay Design



Gene Transcripts

Gene Symbol
NSFL1C
Entrez Gene ID
100513186
Gene Name
NSFL1 cofactor
Gene Aliases
-
Chromosome Location
Chr.17: 33869487 - 33890873 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244443.1 NP_001231372.1 3 - 4 325 69
GenBank mRNA AK232184.1 - 3 - 4 306 69
AK392495.1 - 3 - 4 295 69
DB783818.1 - 3 - 4 325 69

View Full Product Details