Assay Design



Gene Transcripts

Gene Symbol
SERTAD3
Entrez Gene ID
100625125
Gene Name
SERTA domain containing 3
Gene Aliases
-
Chromosome Location
Chr.6: 48709064 - 48713113 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244464.1 NP_001231393.1 1 - 2 155 69
GenBank mRNA AK232391.1 - 1 - 2 89 69
AK240432.1 - 1 - 2 155 69

View Full Product Details