Assay Design



Gene Transcripts

Gene Symbol
BZW1
Entrez Gene ID
100152909
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
-
Chromosome Location
Chr.15: 104525900 - 104543795 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244493.1 NP_001231422.1 11 - 12 1314 109
XM_021074514.1 XP_020930173.1 - 1640 109
GenBank mRNA AK239661.1 - 11 - 12 1320 109
AK343867.1 - 11 - 12 1365 108
AK345146.1 - 5 - 6 912 109
AK394609.1 - - 816 109
AY609907.1 - 11 - 12 1318 109

View Full Product Details