Assay Design



Gene Transcripts

Gene Symbol
SFT2D1
Entrez Gene ID
100513321
Gene Name
SFT2 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 2576815 - 2593516 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244534.1 NP_001231463.1 5 - 6 431 91
GenBank mRNA AK234386.1 - 5 - 6 419 91
AK392532.1 - 5 - 6 408 91
AK400085.1 - 5 - 6 434 91
EW622209.2 - 5 - 6 444 91

View Full Product Details