Assay Design



Gene Transcripts

Gene Symbol
RTN1
Entrez Gene ID
100152394
Gene Name
reticulon 1
Gene Aliases
-
Chromosome Location
Chr.1: 188704260 - 188925855 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244640.1 NP_001231569.1 4 - 5 1954 98
NM_001244641.1 NP_001231570.1 2 - 3 381 98
GenBank mRNA AK391600.1 - 2 - 3 378 98
AK392498.1 - 2 - 3 358 98
AK399868.1 - 2 - 3 378 98

View Full Product Details