Assay Design



Gene Transcripts

Gene Symbol
USB1
Entrez Gene ID
100513409
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
Gene Aliases
C16orf57, C6H16orf57
Chromosome Location
Chr.6: 19852020 - 19874016 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244803.1 NP_001231732.1 6 - 7 774 85
GenBank mRNA AK237059.1 - 6 - 7 774 85

View Full Product Details