Assay Design
Gene Transcripts
- Gene Symbol
- TLR4
- Entrez Gene ID
-
399541
- Gene Name
- toll like receptor 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 258044610 - 258054641 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120680.1 |
|
2 - 3 |
505 |
93 |
GenBank mRNA |
AK230475.1 |
- |
2 - 3 |
351 |
93 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
astrotactin 2 |
ASTN2 |
|
-308.1kb
|
Anti-sense |
Chr. - 256825466 - 257740676 on Build Sscrofa11.1 |
Downstream |
uncharacterized LOC110257084 |
LOC110257084 |
|
252.4kb
|
Anti-sense |
Chr. - 258307056 - 258362548 on Build Sscrofa11.1 |
View Full Product Details