Assay Design
Gene Transcripts
- Gene Symbol
- PVVSPMS
- Entrez Gene ID
-
100232840
- Gene Name
- spermine synthase
- Gene Aliases
-
GSVIVT00006512001
- Chromosome Location
-
Chr.Un_NW_002239346: 688653 - 696170 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB240539.1 |
- |
3 - 4 |
239 |
74 |
View Full Product Details