Assay Design
Gene Transcripts
- Gene Symbol
- VVSUC12
- Entrez Gene ID
-
100232845
- Gene Name
- sucrose transporter-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1_NW_002238162: 1414276 - 1424354 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF021809.1 |
- |
7 - 8 |
755 |
82 |
View Full Product Details