Assay Design
Gene Transcripts
- Gene Symbol
- PDC1
- Entrez Gene ID
-
100232858
- Gene Name
- pyruvate decarboxylase 1
- Gene Aliases
-
GSVIVT00001355001
- Chromosome Location
-
Chr.8: 7060810 - 7063804 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF195868.1 |
- |
3 - 4 |
1246 |
82 |
View Full Product Details