Assay Design
Gene Transcripts
- Gene Symbol
- GRIP22
- Entrez Gene ID
-
100233091
- Gene Name
- ripening-related protein-like
- Gene Aliases
-
GSVIVT00024571001
- Chromosome Location
-
Chr.6: 3064384 - 3065366 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ237989.1 |
- |
1 - 1 |
252 |
91 |
View Full Product Details