Assay Design
Gene Transcripts
- Gene Symbol
- MAP3K
- Entrez Gene ID
-
100232947
- Gene Name
- mitogen-activated protein kinase kinase kinase
- Gene Aliases
-
GSVIVT01017915001
- Chromosome Location
-
Chr.5: 4510826 - 4524541 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY461710.1 |
- |
7 - 8 |
762 |
94 |
View Full Product Details