Assay Design
Gene Transcripts
- Gene Symbol
- HT7
- Entrez Gene ID
-
100232971
- Gene Name
- hexose transporter 7
- Gene Aliases
-
GSVIVT00005628001, HT3
- Chromosome Location
-
Chr.Un_NW_002239434: 457624 - 461651 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY538259.1 |
- |
2 - 3 |
537 |
73 |
AY854146.1 |
- |
2 - 3 |
611 |
73 |
View Full Product Details