Assay Design
Gene Transcripts
- Gene Symbol
- GER6
- Entrez Gene ID
-
100233095
- Gene Name
- germin-like protein 6
- Gene Aliases
-
GSVIVT00016457001
- Chromosome Location
-
Chr.11: 943371 - 944450 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EF064174.1 |
- |
1 - 1 |
550 |
89 |
View Full Product Details