Assay Design
Gene Transcripts
- Gene Symbol
- MYB14
- Entrez Gene ID
-
100233135
- Gene Name
- R2R3 Myb14 transcription factor
- Gene Aliases
-
GSVIVT00028596001
- Chromosome Location
-
Chr.7: 6618114 - 6620357 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EU181424.1 |
- |
1 - 2 |
137 |
73 |
View Full Product Details