Assay Design
Gene Transcripts
- Gene Symbol
- MYB60
- Entrez Gene ID
-
100233072
- Gene Name
- R2R3 MYB60 transcription factor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 1265357 - 1266809 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EU816358.1 |
- |
2 - 3 |
261 |
87 |
View Full Product Details