Assay Design
Gene Transcripts
- Gene Symbol
- AP2
- Entrez Gene ID
-
100268051
- Gene Name
- transcription factor APETALA2
- Gene Aliases
-
GSVIVT00019538001
- Chromosome Location
-
Chr.7: 11563338 - 11567106 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FJ809943.1 |
- |
3 - 4 |
847 |
64 |
View Full Product Details