Assay Design
Gene Transcripts
- Gene Symbol
- CBL03
- Entrez Gene ID
-
100262221
- Gene Name
- calcineurin B-like protein 03
- Gene Aliases
-
VITISV_029390
- Chromosome Location
-
Chr.Un_NW_002240558: 24448 - 32544 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FJ901269.1 |
- |
3 - 4 |
268 |
95 |
View Full Product Details