Assay Design



Gene Transcripts

Gene Symbol
cstf1
Entrez Gene ID
407896
Gene Name
cleavage stimulation factor, 3' pre-RNA, subunit 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683441: 339604 - 346430 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001220.1 NP_001001220.1 4 - 5 799 66
GenBank mRNA BC067934.1 - 4 - 5 799 66
CT030488.1 - 3 - 4 663 66

View Full Product Details