Assay Design



Gene Transcripts

Gene Symbol
sall4
Entrez Gene ID
407860
Gene Name
spalt-like transcription factor 4
Gene Aliases
drrs, hsal4, znf797
Chromosome Location
Chr.10: 27032603 - 27043529 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001458.1 NP_001001458.1 1 - 2 72 86
GenBank mRNA BC067314.1 - 1 - 2 72 86
CT025472.2 - 1 - 2 135 86

View Full Product Details