Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004791.1 | NP_001004791.1 | 5 - 6 | 536 | 94 |
XM_012956698.2 | XP_012812152.1 | 5 - 6 | 657 | 94 | |
GenBank mRNA | BC074526.1 | - | 5 - 6 | 536 | 94 |
CR762157.2 | - | 5 - 6 | 507 | 94 |