Assay Design



Gene Transcripts

Gene Symbol
srsf9
Entrez Gene ID
448015
Gene Name
serine/arginine-rich splicing factor 9
Gene Aliases
sfrs9, srp30c
Chromosome Location
Chr.1: 143179072 - 143183627 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004795.1 NP_001004795.1 1 - 2 330 64
GenBank mRNA BC074531.1 - 1 - 2 330 64
CR760888.2 - 1 - 2 202 64

View Full Product Details