Assay Design



Gene Transcripts

Gene Symbol
mrps34
Entrez Gene ID
448272
Gene Name
mitochondrial ribosomal protein S34
Gene Aliases
-
Chromosome Location
Chr.9: 15724140 - 15728541 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004910.2 NP_001004910.2 1 - 2 185 76
GenBank mRNA BC075342.1 - 1 - 2 166 76
CN120975.1 - 1 - 2 186 76

View Full Product Details