Assay Design



Gene Transcripts

Gene Symbol
pinx1
Entrez Gene ID
448337
Gene Name
PIN2 (TERF1) interacting telomerase inhibitor 1
Gene Aliases
Gno1, Pxr1, lptl, lpts
Chromosome Location
Chr.Un|NW_016683369: 289679 - 361559 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004938.1 NP_001004938.1 2 - 3 185 100
XM_012964033.2 XP_012819487.1 2 - 3 228 100
GenBank mRNA BC075416.1 - 2 - 3 185 100
CR761068.2 - 2 - 3 160 100
CT485754.1 - - 229 100
EU520259.1 - 2 - 3 132 100

View Full Product Details