Assay Design



Gene Transcripts

Gene Symbol
rell2
Entrez Gene ID
448340
Gene Name
RELT-like 2
Gene Aliases
-
Chromosome Location
Chr.3: 18749167 - 18774669 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247979.1 NP_001234908.1 6 - 7 1074 94
NM_001247980.1 NP_001234909.1 6 - 7 1038 94
NR_045263.1 6 - 7 1350 94
GenBank mRNA BC075421.1 - 6 - 7 1342 94
CN075854.1 - 2 - 3 466 94
EL818547.1 - 2 - 3 527 94
ES680851.1 - 2 - 3 523 94

View Full Product Details