Assay Design



Gene Transcripts

Gene Symbol
ints13
Entrez Gene ID
448436
Gene Name
integrator complex subunit 13
Gene Aliases
asun, c12orf11, mat89bb, net48
Chromosome Location
Chr.Un|NW_016685780: 123 - 10398 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004984.1 NP_001004984.1 - 973 63
GenBank mRNA BC075538.1 - - 973 63
CR761707.2 - - 900 63

View Full Product Details