Assay Design



Gene Transcripts

Gene Symbol
ppil1
Entrez Gene ID
448493
Gene Name
peptidylprolyl isomerase like 1
Gene Aliases
-
Chromosome Location
Chr.2: 76611139 - 76614051 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005007.1 NP_001005007.1 2 - 3 273 107
GenBank mRNA BC076653.1 - 2 - 3 273 107

View Full Product Details