Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011053.1 | NP_001011053.1 | 4 - 5 | 400 | 64 |
XM_018095361.1 | XP_017950850.1 | 4 - 5 | 338 | 64 | |
GenBank mRNA | BC084178.1 | - | 4 - 5 | 400 | 64 |
CT485778.1 | - | 4 - 5 | 411 | 64 |