Assay Design
Gene Transcripts
- Gene Symbol
- fuca2
- Entrez Gene ID
-
496744
- Gene Name
- alpha-L-fucosidase 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 67422242 - 67459679 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC088008.1 |
- |
3 - 4 |
818 |
93 |
View Full Product Details