Assay Design



Gene Transcripts

Gene Symbol
pnhd
Entrez Gene ID
394459
Gene Name
pinhead
Gene Aliases
-
Chromosome Location
Chr.1: 85454509 - 85462203 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203534.1 NP_988865.1 2 - 3 193 84
GenBank mRNA AY333424.1 - 2 - 3 193 84
BC170803.1 - 2 - 3 184 84
BC170805.1 - 2 - 3 184 84
CR926370.2 - 2 - 3 169 84

View Full Product Details