Assay Design
Gene Transcripts
- Gene Symbol
- ppia
- Entrez Gene ID
-
394470
- Gene Name
- peptidylprolyl isomerase A (cyclophilin A)
- Gene Aliases
-
CYBP, cyp7, cypa, cyph
- Chromosome Location
-
Chr.3: 107974300 - 107980257 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203544.1 |
NP_988875.1 |
3 - 4 |
273 |
66 |
GenBank mRNA |
BC059741.1 |
- |
3 - 4 |
273 |
66 |
- Gene Symbol
- LOC100488106
- Entrez Gene ID
-
100488106
- Gene Name
- peptidyl-prolyl cis-trans isomerase-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 68591977 - 68592455 on Build UCB_Xtro_10.0
View Full Product Details