Assay Design
Gene Transcripts
- Gene Symbol
- bcap31
- Entrez Gene ID
-
394579
- Gene Name
- B cell receptor associated protein 31
- Gene Aliases
-
6c6-ag, bap31, cdm, dxs1357e
- Chromosome Location
-
Chr.8: 3214079 - 3228395 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203651.1 |
NP_988982.1 |
6 - 7 |
698 |
90 |
GenBank mRNA |
BC061339.1 |
- |
6 - 7 |
698 |
90 |
View Full Product Details