Assay Design



Gene Transcripts

Gene Symbol
tmem38a
Entrez Gene ID
394592
Gene Name
transmembrane protein 38A
Gene Aliases
duf714
Chromosome Location
Chr.1: 100040895 - 100053878 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203665.1 NP_988996.1 4 - 5 606 94
GenBank mRNA BC061269.1 - 4 - 5 606 94
BC122046.1 - 4 - 5 594 94
CU075322.1 - 4 - 5 574 94

View Full Product Details