Assay Design



Gene Transcripts

Gene Symbol
rbm24
Entrez Gene ID
394612
Gene Name
RNA binding motif protein 24
Gene Aliases
SEB4-LIKE, rbm24-a, rbm24-b, rnpc1, rnpc6, seb-4, seb4, seb4-a, xseb4
Chromosome Location
Chr.6: 48593590 - 48604376 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203685.1 NP_989016.1 1 - 2 391 82
GenBank mRNA BC061322.1 - 1 - 2 391 82
CT030513.1 - 1 - 2 400 82

View Full Product Details