Assay Design



Gene Transcripts

Gene Symbol
atp1b2
Entrez Gene ID
394667
Gene Name
ATPase, Na+/K+ transporting, beta 2 polypeptide
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683650: 31804 - 39097 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203739.1 NP_989070.1 4 - 5 656 74
GenBank mRNA BC061283.1 - 4 - 5 672 74
CR760087.2 - 4 - 5 639 74

View Full Product Details