Assay Design



Gene Transcripts

Gene Symbol
psma7
Entrez Gene ID
394668
Gene Name
proteasome subunit alpha 7
Gene Aliases
hspc, psma7-a, psma7-b, rc6-1, xapc7
Chromosome Location
Chr.10: 1136370 - 1142436 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203740.1 NP_989071.1 2 - 3 280 95
GenBank mRNA BC061282.1 - 2 - 3 280 95
CR760155.2 - 2 - 3 255 95

View Full Product Details