Assay Design
Gene Transcripts
- Gene Symbol
- dcaf7
- Entrez Gene ID
-
394701
- Gene Name
- DDB1 and CUL4 associated factor 7
- Gene Aliases
-
wdr68
- Chromosome Location
-
Chr.10: 7924157 - 7930843 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203766.1 |
NP_989097.1 |
4 - 5 |
594 |
64 |
GenBank mRNA |
BC062486.1 |
- |
4 - 5 |
594 |
64 |
View Full Product Details