Assay Design
Gene Transcripts
- Gene Symbol
- sec22b
- Entrez Gene ID
-
394761
- Gene Name
- uncharacterized LOC394761
- Gene Aliases
-
ers-24, sec22b-a, sec22b-b, sec22l1
- Chromosome Location
-
Chr.4: 107712846 - 107721621 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203825.1 |
NP_989156.1 |
3 - 4 |
368 |
72 |
GenBank mRNA |
BC061616.1 |
- |
3 - 4 |
368 |
72 |
View Full Product Details