Assay Design



Gene Transcripts

Gene Symbol
tax1bp3
Entrez Gene ID
394838
Gene Name
Tax1 binding protein 3
Gene Aliases
tip-1
Chromosome Location
Chr.2: 31870833 - 31883254 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203899.1 NP_989230.1 2 - 3 174 69
GenBank mRNA BC063221.1 - 2 - 3 174 69
BC080496.1 - 2 - 3 171 69
CR855715.2 - 2 - 3 195 69

View Full Product Details