Assay Design
Gene Transcripts
- Gene Symbol
- cndp2
- Entrez Gene ID
-
394907
- Gene Name
- CNDP dipeptidase 2 (metallopeptidase M20 family)
- Gene Aliases
-
cn2, cpgl, darmin-r
- Chromosome Location
-
Chr.Un|NW_016683375: 1670074 - 1684505 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203958.1 |
NP_989289.1 |
10 - 11 |
1438 |
64 |
GenBank mRNA |
BC064175.1 |
- |
10 - 11 |
1438 |
64 |
View Full Product Details