Assay Design



Gene Transcripts

Gene Symbol
lamtor1
Entrez Gene ID
394922
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
Gene Aliases
c11orf59, p27rf-rho, pdro, ragulator1
Chromosome Location
Chr.Un|NW_016685432: 988 - 4116 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203972.1 NP_989303.1 1 - 2 293 76
GenBank mRNA BC064269.1 - 1 - 2 293 76
CT025450.2 - 1 - 2 260 76

View Full Product Details