Assay Design



Gene Transcripts

Gene Symbol
srsf2
Entrez Gene ID
394953
Gene Name
serine/arginine-rich splicing factor 2
Gene Aliases
pr264, sc-35, sc35, sfrs2, sfrs2a, srp30b
Chromosome Location
Chr.10: 36954984 - 36958856 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203997.1 NP_989328.1 1 - 2 481 86
GenBank mRNA BC064167.1 - 1 - 2 481 86
CR760874.2 - 1 - 2 389 86

View Full Product Details