Assay Design



Gene Transcripts

Gene Symbol
ppp3r1
Entrez Gene ID
395037
Gene Name
protein phosphatase 3 regulatory subunit B, alpha
Gene Aliases
-
Chromosome Location
Chr.5: 30906249 - 30930146 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204069.1 NP_989400.1 4 - 5 438 75
GenBank mRNA AY336972.1 - 4 - 5 439 75
BC064854.1 - 4 - 5 438 75

View Full Product Details