Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_204073.1 | NP_989404.1 | 3 - 4 | 358 | 90 |
XM_018091892.1 | XP_017947381.1 | 3 - 4 | 418 | 90 | |
GenBank mRNA | BC064864.1 | - | 3 - 4 | 358 | 90 |
CR855787.2 | - | 3 - 4 | 353 | 90 |