Assay Design



Gene Transcripts

Gene Symbol
mrto4
Entrez Gene ID
448696
Gene Name
MRT4 homolog, ribosome maturation factor
Gene Aliases
mrt4
Chromosome Location
Chr.Un|NW_016683515: 300877 - 304130 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005116.2 NP_001005116.1 - 588 108
GenBank mRNA BC077055.1 - - 590 108
CR760103.2 - - 579 108

View Full Product Details