Assay Design



Gene Transcripts

Gene Symbol
cops9
Entrez Gene ID
448732
Gene Name
COP9 signalosome subunit 9
Gene Aliases
myeov2
Chromosome Location
Chr.5: 123872861 - 123876044 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005143.1 NP_001005143.1 2 - 3 226 106
GenBank mRNA BC077691.1 - 2 - 3 226 106
CR761705.2 - 2 - 3 161 106

View Full Product Details